Assay Details
Gene Symbol: Cfap100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004854153.2 | XP_004854210.1 | 10 - 11 | 1185 | 67 | |
XM_021265998.1 | XP_021121657.1 | 9 - 10 | 1242 | 67 | ||
XM_021265999.1 | XP_021121658.1 | 9 - 10 | 1288 | 67 | ||
XM_021266002.1 | XP_021121661.1 | 10 - 11 | 1314 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
