Assay Details
Gene Symbol: Hsd17b12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004851999.3 | XP_004852056.1 | 8 - 9 | 713 | 69 | |
XM_021264326.1 | XP_021119985.1 | 8 - 9 | 723 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
