Assay Details
| Gene Symbol: Atp1b3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004834365.2 | XP_004834422.1 | 4 - 5 | 689 | 76 | |
| XM_004834366.2 | XP_004834423.1 | 6 - 7 | 851 | 76 | ||
More Information
Set Membership: |
|

Atp1b3
Genomic Map