Assay Details
Gene Symbol: Ddx50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004846173.2 | XP_004846230.1 | 13 - 14 | 2042 | 70 | |
XM_004846174.2 | XP_004846231.1 | 12 - 13 | 1881 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
