Assay Details
| Gene Symbol: Atp6v1g1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004849497.3 | XP_004849554.1 | 1 - 2 | 211 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atp6v1g1| Gene Symbol: Atp6v1g1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004849497.3 | XP_004849554.1 | 1 - 2 | 211 | 78 | |
Set Membership: |
|