Assay Details
Gene Symbol: Akirin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004860228.3 | XP_004860285.1 | 4 - 5 | 728 | 83 | |
XM_021240785.1 | XP_021096444.1 | 4 - 5 | 522 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
