Assay Details
Gene Symbol: Tmem9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004851395.3 | XP_004851452.1 | 4 - 5 | 928 | 66 | |
XM_013074609.2 | XP_012930063.1 | 3 - 4 | 435 | 66 | ||
XM_021263763.1 | XP_021119422.1 | 4 - 5 | 849 | 66 | ||
XM_021263764.1 | XP_021119423.1 | 4 - 5 | 500 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
