Assay Details
Gene Symbol: LOC101723971 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013068049.2 | XP_012923503.1 | 2 - 3 | 293 | 67 | |
XM_013068050.2 | XP_012923504.1 | 2 - 3 | 321 | 67 | ||
XM_013068051.2 | XP_012923505.1 | 2 - 3 | 293 | 67 | ||
XM_021249559.1 | XP_021105218.1 | 1 - 2 | 571 | 67 | ||
XM_021249560.1 | XP_021105219.1 | 2 - 3 | 318 | 67 | ||
XM_021249561.1 | XP_021105220.1 | 2 - 3 | 289 | 67 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013068049.2, XM_013068050.2, XM_013068051.2, XM_021249559.1, XM_021249560.1, XM_021249561.1 transcripts.
Set Membership: |
![]() ![]() |
