Assay Details
Gene Symbol: LOC101723971 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013068049.2 | XP_012923503.1 | 5 - 6 | 875 | 99 | |
XM_013068050.2 | XP_012923504.1 | 5 - 6 | 903 | 99 | ||
XM_013068051.2 | XP_012923505.1 | 5 - 6 | 875 | 99 | ||
XM_021249551.1 | XP_021105210.1 | 6 - 7 | 886 | 99 | ||
XM_021249552.1 | XP_021105211.1 | 6 - 7 | 917 | 99 | ||
XM_021249553.1 | XP_021105212.1 | 6 - 7 | 914 | 99 | ||
XM_021249554.1 | XP_021105213.1 | 6 - 7 | 886 | 99 | ||
XM_021249555.1 | XP_021105214.1 | 6 - 7 | 889 | 99 | ||
XM_021249556.1 | XP_021105215.1 | 6 - 7 | 827 | 99 | ||
XM_021249557.1 | XP_021105216.1 | 6 - 7 | 882 | 99 | ||
XM_021249558.1 | XP_021105217.1 | 6 - 7 | 779 | 99 | ||
XM_021249559.1 | XP_021105218.1 | 4 - 5 | 1153 | 99 | ||
XM_021249560.1 | XP_021105219.1 | 5 - 6 | 900 | 99 | ||
XM_021249561.1 | XP_021105220.1 | 5 - 6 | 871 | 99 | ||
XM_021249562.1 | XP_021105221.1 | 5 - 6 | 1104 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() |
