Assay Details
Gene Symbol: LOC101723971 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013068049.2 | XP_012923503.1 | 7 - 8 | 1146 | 103 | |
XM_013068050.2 | XP_012923504.1 | 7 - 8 | 1174 | 103 | ||
XM_013068051.2 | XP_012923505.1 | 7 - 8 | 1146 | 103 | ||
XM_021249551.1 | XP_021105210.1 | 8 - 9 | 1157 | 103 | ||
XM_021249552.1 | XP_021105211.1 | 8 - 9 | 1188 | 103 | ||
XM_021249553.1 | XP_021105212.1 | 8 - 9 | 1185 | 103 | ||
XM_021249554.1 | XP_021105213.1 | 8 - 9 | 1157 | 103 | ||
XM_021249555.1 | XP_021105214.1 | 8 - 9 | 1160 | 103 | ||
XM_021249556.1 | XP_021105215.1 | 8 - 9 | 1098 | 103 | ||
XM_021249557.1 | XP_021105216.1 | 8 - 9 | 1153 | 103 | ||
XM_021249558.1 | XP_021105217.1 | 8 - 9 | 1050 | 103 | ||
XM_021249559.1 | XP_021105218.1 | 6 - 7 | 1424 | 103 | ||
XM_021249560.1 | XP_021105219.1 | 7 - 8 | 1171 | 103 | ||
XM_021249561.1 | XP_021105220.1 | 7 - 8 | 1142 | 103 | ||
XM_021249562.1 | XP_021105221.1 | 7 - 8 | 1375 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() |
