Assay Details
| Gene Symbol: Ctsc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004868209.3 | XP_004868266.1 | 1 - 2 | 273 | 63 | |
| XM_004868211.3 | XP_004868268.1 | 1 - 2 | 281 | 63 | ||
| XM_004868212.3 | XP_004868269.1 | 1 - 2 | 280 | 63 | ||
| XM_004868213.3 | XP_004868270.1 | 1 - 2 | 280 | 63 | ||
More Information
Set Membership: |
|

Ctsc
Genomic Map