Assay Details
Gene Symbol: Jkamp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004837456.3 | XP_004837513.1 | 4 - 5 | 603 | 77 | |
XM_004837457.3 | XP_004837514.1 | 4 - 5 | 551 | 77 | ||
XM_004837458.3 | XP_004837515.1 | 4 - 5 | 603 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
