Assay Details
| Gene Symbol: Setd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004836977.3 | XP_004837034.1 | 10 - 11 | 1121 | 64 | |
| XM_004836978.2 | XP_004837035.1 | 9 - 10 | 1101 | 64 | ||
| XM_004836979.2 | XP_004837036.1 | 9 - 10 | 1026 | 64 | ||
| XM_004836980.3 | XP_004837037.1 | 10 - 11 | 1107 | 64 | ||
| XM_004836981.3 | XP_004837038.1 | 10 - 11 | 1123 | 64 | ||
| XM_021244481.1 | XP_021100140.1 | 9 - 10 | 1011 | 64 | ||
| XM_021244485.1 | XP_021100144.1 | 9 - 10 | 1086 | 64 | ||
More Information
Set Membership: |
|

Setd3
Genomic Map