Assay Details
Gene Symbol: Nsd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004855349.3 | XP_004855406.1 | 17 - 18 | 3559 | 103 | |
XM_004855350.3 | XP_004855407.1 | 18 - 19 | 3663 | 103 | ||
XM_004855352.3 | XP_004855409.1 | 17 - 18 | 3559 | 103 | ||
XM_013076069.2 | XP_012931523.1 | 16 - 17 | 3412 | 103 |

More Information
Set Membership: |
![]() ![]() |
