Assay Details
Gene Symbol: Nsd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004855349.3 | XP_004855406.1 | 21 - 22 | 4205 | 73 | |
XM_004855350.3 | XP_004855407.1 | 22 - 23 | 4309 | 73 | ||
XM_004855352.3 | XP_004855409.1 | 21 - 22 | 4172 | 73 | ||
XM_013076069.2 | XP_012931523.1 | 20 - 21 | 4058 | 73 |
