Assay Details
Gene Symbol: Nsd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004855349.3 | XP_004855406.1 | 8 - 9 | 2245 | 68 | |
XM_004855350.3 | XP_004855407.1 | 9 - 10 | 2349 | 68 | ||
XM_004855352.3 | XP_004855409.1 | 8 - 9 | 2245 | 68 | ||
XM_004855354.3 | XP_004855411.1 | 8 - 9 | 2214 | 68 | ||
XM_013076069.2 | XP_012931523.1 | 8 - 9 | 2245 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
