Assay Details
Gene Symbol: Nck1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004834296.3 | XP_004834353.1 | 4 - 5 | 1181 | 124 | |
XM_004834297.3 | XP_004834354.1 | 5 - 6 | 1252 | 124 | ||
XM_004834298.3 | XP_004834355.1 | 4 - 5 | 1188 | 124 | ||
XM_004834299.3 | XP_004834356.1 | 3 - 4 | 1118 | 124 | ||
XM_021247354.1 | XP_021103013.1 | 8 - 9 | 1645 | 124 | ||
XM_021247396.1 | XP_021103055.1 | 6 - 7 | 1437 | 124 | ||
XM_021247440.1 | XP_021103099.1 | 7 - 8 | 1570 | 124 | ||
XM_021247470.1 | XP_021103129.1 | 7 - 8 | 1574 | 124 | ||
XM_021247560.1 | XP_021103219.1 | 5 - 6 | 1373 | 124 | ||
XM_021247597.1 | XP_021103256.1 | 7 - 8 | 1581 | 124 | ||
XM_021247625.1 | XP_021103284.1 | 6 - 7 | 1506 | 124 | ||
XM_021247698.1 | XP_021103357.1 | 2 - 3 | 804 | 124 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
