Assay Details
Gene Symbol: Prr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004845417.3 | XP_004845474.1 | 4 - 5 | 585 | 58 | |
XM_013072543.2 | XP_012927997.1 | 5 - 6 | 488 | 58 | ||
XM_013072544.2 | XP_012927998.1 | 5 - 6 | 488 | 58 | ||
XM_013072545.2 | XP_012927999.1 | 5 - 6 | 411 | 58 | ||
XM_013072547.2 | XP_012928001.1 | 4 - 5 | 331 | 58 | ||
XM_021258817.1 | XP_021114476.1 | 5 - 6 | 488 | 58 | ||
XM_021258819.1 | XP_021114478.1 | 5 - 6 | 488 | 58 | ||
XM_021258820.1 | XP_021114479.1 | 4 - 5 | 586 | 58 | ||
XM_021258821.1 | XP_021114480.1 | 3 - 4 | 505 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() |
