Assay Details
Gene Symbol: LOC101703804 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004864580.2 | XP_004864637.1 | 2 - 3 | 579 | 80 | |
XM_004864582.2 | XP_004864639.1 | 2 - 3 | 548 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() |
