Assay Details
Gene Symbol: LOC101703804 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004864580.2 | XP_004864637.1 | 7 - 8 | 1261 | 97 | |
XM_004864582.2 | XP_004864639.1 | 7 - 8 | 1230 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
