Assay Details
| Gene Symbol: Glrb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004868544.3 | XP_004868601.1 | 8 - 9 | 1040 | 78 | |
| XM_004868545.3 | XP_004868602.1 | 7 - 8 | 1070 | 78 | ||
| XM_021247454.1 | XP_021103113.1 | 7 - 8 | 903 | 78 | ||
| XM_021247455.1 | XP_021103114.1 | 7 - 8 | 945 | 78 | ||
More Information
Set Membership: |
|

Glrb
Genomic Map