Assay Details
| Gene Symbol: Nectin3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004835753.3 | XP_004835810.1 | 2 - 3 | 675 | 102 | |
| XM_004835754.3 | XP_004835811.1 | 2 - 3 | 674 | 102 | ||
| XM_004835755.3 | XP_004835812.1 | 2 - 3 | 677 | 102 | ||
| XM_021263918.1 | XP_021119577.1 | 2 - 3 | 681 | 102 | ||
| XM_021263922.1 | XP_021119581.1 | 2 - 3 | 677 | 102 | ||
| XM_021263931.1 | XP_021119590.1 | 2 - 3 | 677 | 102 | ||
| XM_021263937.1 | XP_021119596.1 | 2 - 3 | 674 | 102 | ||
More Information
Set Membership: |
|

Nectin3
Genomic Map