Assay Details
Gene Symbol: Nectin3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004835753.3 | XP_004835810.1 | 3 - 4 | 973 | 76 | |
XM_004835754.3 | XP_004835811.1 | 3 - 4 | 972 | 76 | ||
XM_004835755.3 | XP_004835812.1 | 3 - 4 | 975 | 76 | ||
XM_021263918.1 | XP_021119577.1 | 3 - 4 | 979 | 76 | ||
XM_021263922.1 | XP_021119581.1 | 3 - 4 | 975 | 76 | ||
XM_021263931.1 | XP_021119590.1 | 3 - 4 | 975 | 76 | ||
XM_021263937.1 | XP_021119596.1 | 3 - 4 | 972 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
