Assay Details
Gene Symbol: Kirrel1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004858768.3 | XP_004858825.1 | 14 - 15 | 2098 | 76 | |
XM_021239852.1 | XP_021095511.1 | 14 - 15 | 2333 | 76 | ||
XM_021239853.1 | XP_021095512.1 | 14 - 15 | 2285 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
