Assay Details
| Gene Symbol: Kirrel1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004858768.3 | XP_004858825.1 | 5 - 6 | 908 | 67 | |
| XM_021239852.1 | XP_021095511.1 | 5 - 6 | 1143 | 67 | ||
| XM_021239853.1 | XP_021095512.1 | 5 - 6 | 1143 | 67 | ||
More Information
Set Membership: |
|

Kirrel1
Genomic Map