Assay Details
Gene Symbol: Ccdc106 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004866615.3 | XP_004866672.1 | 4 - 5 | 684 | 72 | |
XM_013066337.2 | XP_012921791.1 | 4 - 5 | 868 | 72 | ||
XM_013066338.2 | XP_012921792.1 | 4 - 5 | 705 | 72 | ||
XM_021245946.1 | XP_021101605.1 | 4 - 5 | 709 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
