Assay Details
Gene Symbol: Rsg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004850363.2 | XP_004850420.1 | 3 - 4 | 380 | 60 | |
XM_013074365.2 | XP_012929819.1 | 4 - 5 | 1081 | 60 | ||
XM_021262868.1 | XP_021118527.1 | 4 - 5 | 1057 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
