Assay Details
Gene Symbol: Nutf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004843008.3 | XP_004843065.1 | 3 - 4 | 491 | 67 | |
XM_004843009.2 | XP_004843066.1 | 3 - 4 | 396 | 67 | ||
XM_004843010.3 | XP_004843067.1 | 3 - 4 | 331 | 67 | ||
XM_004843012.3 | XP_004843069.1 | 4 - 5 | 427 | 67 | ||
XM_021256841.1 | XP_021112500.1 | 3 - 4 | 339 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
