Assay Details
Gene Symbol: Ptp4a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004837877.3 | XP_004837934.1 | 4 - 5 | 1230 | 56 | |
XM_004837878.3 | XP_004837935.1 | 4 - 5 | 1320 | 56 | ||
XM_004837879.3 | XP_004837936.1 | 4 - 5 | 1277 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
