Assay Details
Gene Symbol: Eif4a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004834818.3 | XP_004834875.1 | 8 - 9 | 1064 | 61 | |
XM_021255886.1 | XP_021111545.1 | 7 - 8 | 872 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
