Assay Details
| Gene Symbol: Hacd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004855613.2 | XP_004855670.1 | 9 - 10 | 1057 | 80 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hacd3| Gene Symbol: Hacd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004855613.2 | XP_004855670.1 | 9 - 10 | 1057 | 80 | |
Set Membership: |
|