Assay Details
Gene Symbol: Bgn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004874920.3 | XP_004874977.1 | 6 - 7 | 941 | 59 | |
XM_021252330.1 | XP_021107989.1 | 6 - 7 | 810 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
