Assay Details
Gene Symbol: Nsg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004847300.2 | XP_004847357.1 | 3 - 4 | 379 | 63 | |
XM_004847301.2 | XP_004847358.1 | 3 - 4 | 376 | 63 | ||
XM_013073047.2 | XP_012928501.1 | 3 - 4 | 289 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
