Assay Details
| Gene Symbol: LOC101698131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021260358.1 | XP_021116017.1 | 6 - 7 | 820 | 96 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101698131| Gene Symbol: LOC101698131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021260358.1 | XP_021116017.1 | 6 - 7 | 820 | 96 | |
Set Membership: |
|