Assay Details
Gene Symbol: Ufc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004858674.3 | XP_004858731.1 | 4 - 5 | 613 | 65 | |
XM_004858675.3 | XP_004858732.1 | 4 - 5 | 578 | 65 | ||
XM_004858676.3 | XP_004858733.1 | 4 - 5 | 580 | 65 | ||
XM_004858679.3 | XP_004858736.1 | 3 - 4 | 459 | 65 | ||
XM_004858681.3 | XP_004858738.1 | 3 - 4 | 422 | 65 | ||
XM_021239616.1 | XP_021095275.1 | 4 - 5 | 545 | 65 | ||
XM_021239617.1 | XP_021095276.1 | 4 - 5 | 483 | 65 | ||
XM_021239618.1 | XP_021095277.1 | 4 - 5 | 455 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
