Assay Details
Gene Symbol: Atf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004862976.3 | XP_004863033.1 | 6 - 7 | 869 | 72 | |
XM_004862978.3 | XP_004863035.1 | 5 - 6 | 745 | 72 | ||
XM_013079005.2 | XP_012934459.1 | 6 - 7 | 863 | 72 | ||
XM_013079006.2 | XP_012934460.1 | 4 - 5 | 565 | 72 | ||
XM_021243067.1 | XP_021098726.1 | 5 - 6 | 594 | 72 | ||
XM_021243068.1 | XP_021098727.1 | 5 - 6 | 697 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
