Assay Details
Gene Symbol: Dhrs7c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004857521.3 | XP_004857578.1 | 2 - 3 | 309 | 63 | |
XM_004857522.3 | XP_004857579.1 | 2 - 3 | 309 | 63 | ||
XM_021238434.1 | XP_021094093.1 | 2 - 3 | 309 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
