Assay Details
Gene Symbol: Erg28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004837155.3 | XP_004837212.1 | 2 - 3 | 375 | 59 | |
XM_021246257.1 | XP_021101916.1 | 2 - 3 | 377 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
