Assay Details
Gene Symbol: Yeats4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004845062.3 | XP_004845119.1 | 1 - 2 | 342 | 90 | |
XM_013072339.2 | XP_012927793.1 | 1 - 2 | 347 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
