Assay Details
Gene Symbol: Efs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004864162.3 | XP_004864219.1 | 4 - 5 | 1804 | 68 | |
XM_004864164.3 | XP_004864221.1 | 4 - 5 | 2121 | 68 | ||
XM_021243832.1 | XP_021099491.1 | 5 - 6 | 1904 | 68 | ||
XM_021243833.1 | XP_021099492.1 | 5 - 6 | 2217 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
