Assay Details
| Gene Symbol: Klrg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004869538.2 | XP_004869595.1 | 4 - 5 | 1569 | 65 | |
| XM_021248147.1 | XP_021103806.1 | 3 - 4 | 1464 | 65 | ||
More Information
Set Membership: |
|

Klrg1
Genomic Map