Assay Details
| Gene Symbol: Grhpr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004874574.3 | XP_004874631.1 | 2 - 3 | 298 | 65 | |
| XM_004874575.3 | XP_004874632.1 | 2 - 3 | 297 | 65 | ||
| XM_004874576.3 | XP_004874633.1 | 2 - 3 | 298 | 65 | ||
| XM_004874577.3 | XP_004874634.1 | 2 - 3 | 251 | 65 | ||
More Information
Set Membership: |
|

Grhpr
Genomic Map