Assay Details
| Gene Symbol: Asphd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004843342.2 | XP_004843399.1 | 3 - 4 | 1342 | 89 | |
| XM_021257145.1 | XP_021112804.1 | 4 - 5 | 1449 | 89 | ||
More Information
Set Membership: |
|

Asphd2
Genomic Map