Assay Details
Gene Symbol: Nr0b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004850061.3 | XP_004850118.1 | 2 - 3 | 2149 | 92 | |
XM_021262609.1 | XP_021118268.1 | 3 - 4 | 2309 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
