Assay Details
Gene Symbol: Shd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004865761.2 | XP_004865818.1 | 3 - 4 | 983 | 71 | |
XM_021245006.1 | XP_021100665.1 | 3 - 4 | 990 | 71 | ||
XM_021245007.1 | XP_021100666.1 | 3 - 4 | 928 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
