Assay Details
Gene Symbol: Gins2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004842748.2 | XP_004842805.1 | 4 - 5 | 538 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
