Assay Details
Gene Symbol: Sil1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004841824.2 | XP_004841881.1 | 6 - 7 | 665 | 63 | |
XM_004841825.3 | XP_004841882.1 | 5 - 6 | 563 | 63 | ||
XM_021255895.1 | XP_021111554.1 | 5 - 6 | 608 | 63 | ||
XM_021255896.1 | XP_021111555.1 | 5 - 6 | 633 | 63 | ||
XM_021255897.1 | XP_021111556.1 | 5 - 6 | 542 | 63 | ||
XM_021255898.1 | XP_021111557.1 | 5 - 6 | 508 | 63 | ||
XM_021255899.1 | XP_021111558.1 | 5 - 6 | 500 | 63 | ||
XM_021255900.1 | XP_021111559.1 | 5 - 6 | 608 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
