Assay Details
| Gene Symbol: Klf10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004850239.3 | XP_004850296.1 | 3 - 4 | 1285 | 73 | |
| XM_021262744.1 | XP_021118403.1 | 4 - 5 | 1532 | 73 | ||
More Information
Set Membership: |
|

Klf10
Genomic Map