Assay Details
Gene Symbol: Arl13b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004857923.3 | XP_004857980.1 | 2 - 3 | 444 | 65 | |
XM_021238817.1 | XP_021094476.1 | 3 - 4 | 480 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
