Assay Details
Gene Symbol: Abhd14b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004834187.3 | XP_004834244.2 | 2 - 3 | 335 | 53 | |
XM_004834189.2 | XP_004834246.1 | 2 - 3 | 361 | 53 |

More Information
Set Membership: |
![]() ![]() ![]() |
